Skip to Content
Merck
All Photos(2)

Documents

D136808

Sigma-Aldrich

2,2-Dimethoxypropane

reagent grade, 98%

Synonym(s):

Acetone dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OCH3)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
635678
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

3.59 (vs air)

vapor pressure

60 mmHg ( 15.8 °C)

Assay

98%

form

liquid

expl. lim.

31 %, 58 °F
6 %, 27 °F

refractive index

n20/D 1.378 (lit.)

bp

83 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)OC

InChI

1S/C5H12O2/c1-5(2,6-3)7-4/h1-4H3

InChI key

HEWZVZIVELJPQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethoxypropane (DMP) is an organic building block commonly employed as a precursor to generate 2-methoxypropene (MPP). The degradation study of DMP in ionic liquids showed the formation of MPP and 2-ethoxypropene (EPP) in an identical ratio due to the tunneling effect. Conformational analysis of DMP based on ab initio calculations and matrix isolation infrared spectroscopy has been reported. DMP reacts with water to produce methanol and acetone. This reaction has been employed in a method for the quantification of water in natural products by gas-liquid chromatography. Acidified DMP has been employed for the dehydration of biological samples.

Application

Reagent for the preparation of 1,2-diols as acetonides.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greene, T.W. Wuts, P.G.M.
Protective Groups in Organic Synthesis, 123-123 (1991)
Davit Jishkariani et al.
ACS nano, 11(8), 7958-7966 (2017-08-05)
Janus nanoparticles (NPs) often referred to as nanosized analogs of molecular surfactants are amphiphilic structures with potential applications in materials science, biomedicine, and catalysis, and their synthesis and self-assembly into complex architectures remain challenging. Here, we demonstrate the preparation of
Gas chromatographic determination of water in natural products by reaction with 2,2-dimethoxypropane.
Mary NY.
Journal of Pharmaceutical Sciences, 58(9), 1089-1092 (1969)
The importance of tunneling effect to the decomposition of 2, 2-dimethoxypropane and 2,2-diethoxypropane.
Jiang H, et al.
Chemical Physics Letters, 406(4), 489-494 (2005)
H J Beckmann et al.
Histochemistry, 76(3), 407-412 (1982-01-01)
Dehydration with 2,2-dimethoxypropane acidified with either 1 N HCL or 0.05 N HCL in an 1:100 ratio has been found to extract various biologically active lipids. The effect is enhanced by an increased proton concentration and seems to occur only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service