Skip to Content
Merck

Skip To

COD500

COD 500 mg/L Calibration Standard

certified reference material

Synonym(s):

Chemical Oxygen Demand Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COD500-100

concentration

in H2O

technique(s)

TOC: suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

This CRM is intended for the calibration and validation of analytical instruments used to measure chemical oxygen demand (COD).

Features and Benefits

  • Verified and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to the SI and higher order standards from NIST
  • Assigned value based on a purity determination by mass balance and gravimetrically prepared value
  • Homogeneity assessed in accordance with ISO Guide 35
  • Available in high-density polyethylene bottles of volumes 100 and 500 mL for an assured safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
  • Prepared using deionized water as a solvent

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BOD500BOD1000BOD200
application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

technique(s)

TOC: suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

matrix material

format

matrix material

format

matrix material

format

matrix material

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

RTC COD500-100

manufacturer/tradename

RTC BOD500-100

manufacturer/tradename

RTC BOD1000-500

manufacturer/tradename

RTC BOD200-100

concentration

in H2O

concentration

in H2O

concentration

in H2O

concentration

in H2O


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
COD500-100ML04061838092403
COD500-500ML04061838261663

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters