Skip to Content
Merck
All Photos(1)

Documents

C6395

Supelco

Cannabidiol solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Synonym(s):

CBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CCCCCc1cc(O)c([C@@H]2C=C(C)CC[C@H]2C(C)=C)c(O)c1

InChI

1S/C21H30O2/c1-5-6-7-8-16-12-19(22)21(20(23)13-16)18-11-15(4)9-10-17(18)14(2)3/h11-13,17-18,22-23H,2,5-10H2,1,3-4H3/t17-,18+/m0/s1

InChI key

QHMBSVQNZZTUGM-ZWKOTPCHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cannabidiol belongs to the group of active cannabinoids, available in neutral form, derived from Cannabis species and is available as a main component of illicit drugs such as hashish (cannabis resin) and marijuana (herbal cannabis).

Application

Cannabidiol is used as an analytical reference standard for the quantification of the analyte in hashish drug samples and plasma samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous separation and identification of hashish constituents by coupled liquid chromatography-mass spectrometry (HPLC-MS)
Rustichelli.C, et al.
Chromatographia, 43(3-4), 129-134 (1996)
Maiko Tsuchiya et al.
International journal of molecular sciences, 20(24) (2019-12-15)
Bone metabolism is strictly regulated, and impaired regulation caused by hormonal imbalances induces systemic bone loss. Local bone loss caused by tumor invasion into bone is suggested to be induced by the generation of cytokines, which affect bone metabolism, by
Development of a simple and sensitive HPLC?UV method for the simultaneous determination of cannabidiol and ?9-tetrahydrocannabinol in rat plasma
Zgair A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 114(3-4), 145-151 (2015)
Liliana Soroceanu et al.
Cancer research, 73(5), 1559-1569 (2012-12-18)
Glioblastoma is the most common form of primary adult brain tumors. A majority of glioblastomas grow invasively into distant brain tissue, leading to tumor recurrence, which is ultimately incurable. It is, therefore, essential to discover master regulators that control glioblastoma
Alline Cristina Campos et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 367(1607), 3364-3378 (2012-10-31)
Cannabidiol (CBD) is a major phytocannabinoid present in the Cannabis sativa plant. It lacks the psychotomimetic and other psychotropic effects that the main plant compound Δ(9)-tetrahydrocannabinol (THC) being able, on the contrary, to antagonize these effects. This property, together with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service