Skip to Content
Merck
All Photos(1)

Key Documents

BCR297

2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153)

BCR®, certified reference material

Synonym(s):

2,2′,4,4′,5,5′-Hexachlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Beilstein:
1990497
Ballschmiter Number:
153
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

neat

storage temp.

2-8°C

SMILES string

Clc1cc(Cl)c(cc1Cl)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H4Cl6/c13-7-3-11(17)9(15)1-5(7)6-2-10(16)12(18)4-8(6)14/h1-4H

InChI key

MVWHGTYKUMDIHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR297

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Valera et al.
Neurotoxicology, 33(5), 1067-1074 (2012-12-12)
Studies conducted in the Faeroe Islands and Japan suggest a negative impact of mercury on heart rate variability (HRV) among children while the results regarding blood pressure (BP) are less consistent. To assess the impact of mercury on HRV and
Chao Han et al.
Journal of hazardous materials, 248-249, 313-321 (2013-02-19)
Polychlorinated biphenyls (PCBs) possessed much potential hazard to environment because of its chemical stability and biological toxicity. Here, we identified the binding mode of a representative compound, PCB153, to human serum albumin (HSA) using fluorescence and molecular dynamics simulation methods.
Cristina L Quinn et al.
Environment international, 49, 83-91 (2012-09-18)
Concentrations of persistent organic pollutants (POPs) in Inuit populations have been observed to decrease over the last decade. The main objective of this study was to develop a methodology to quantify the potential influence of intergenerational dietary transitions on human
Omar Cauli et al.
Toxicology, 311(1-2), 61-68 (2012-12-12)
Polychlorinated biphenyls (PCBs) and methylmercury (MeHg) are persistent organic pollutants accumulating in the food chain. Pre- and neonatal exposure to these neurotoxicants may affect brain development and lead to long-lasting alterations in cerebral function, which can result in motor alterations
Xue Shi et al.
Journal of proteome research, 11(7), 3805-3815 (2012-06-13)
Polychlorinated biphenyls (PCBs) are persistent organic pollutants and have been associated with abnormal liver enzymes and suspected nonalcoholic fatty liver disease (NAFLD), obesity, and the metabolic syndrome in epidemiological studies. In epidemiological surveys of human PCB exposure, PCB 153 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service