
2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153)

Synonym(s):
2,2′,4,4′,5,5′-Hexachlorobiphenyl
Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Products
Formula Weight
Manufacturer
Agency
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing