Skip to Content
Merck
All Photos(1)

Key Documents

BCR133

Dibenzo[a,e]pyrene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H14
CAS Number:
Molecular Weight:
302.37
Beilstein:
2054070
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)cc3c4ccccc4c5cccc6ccc2c3c56

InChI

1S/C24H14/c1-2-8-17-16(6-1)14-22-19-10-4-3-9-18(19)20-11-5-7-15-12-13-21(17)24(22)23(15)20/h1-14H

InChI key

KGHMWBNEMFNJFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR133

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vahideh Moradi et al.
Iranian journal of pharmaceutical research : IJPR, 19(3), 183-194 (2021-03-09)
This research reports a validated multi-residue method based on gas chromatography coupled mass spectrometry technique for analysis of 24 Polycyclic Aromatic Hydrocarbons (PAHs) in traditional and semi-industrial Taftoon bread using QuEChERS sample preparation. Matrix effect studies were performed by comparing
Anthony Santana et al.
Analytica chimica acta, 1100, 163-173 (2020-01-29)
Polycyclic aromatic hydrocarbons (PAHs) are some of the most common environmental pollutants encountered worldwide. Eco-toxicological studies attribute a significant portion of the biological activity of PAH contaminated samples to the presence of high molecular weight PAHs (HMW-PAHs), i.e. PAHs with
Carmela Lestingi et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(7), 1140-1152 (2017-05-18)
A gas-chromatographic single-quadrupole analytical method for the analysis of the 16 priority European Union (EU) polycyclic aromatic hydrocarbons (PAHs) in food is presented. The method fulfils the request of Regulation EU 836/2011 for an analytical procedure to be used for
P D Devanesan et al.
Chemical research in toxicology, 3(6), 580-586 (1990-11-01)
Dibenzo[a,l]pyrene (DB[a,l]P) is one of the most potent carcinogens ever tested in mouse skin and rat mammary gland. DB[a,l]P is present in cigarette smoke and, presumably, in other environmental pollutants. Metabolism and mutagenicity studies of this compound compared to the
Dibenzo[a,e]pyrene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 327-330 (1983-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service