Skip to Content
Merck
All Photos(1)

Documents

196231

Sigma-Aldrich

4-Phenylcyclohexanone

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H9(=O)
CAS Number:
Molecular Weight:
174.24
Beilstein:
2045904
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

mp

73-77 °C (lit.)

SMILES string

O=C1CCC(CC1)c2ccccc2

InChI

1S/C12H14O/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-5,11H,6-9H2

InChI key

YKAYMASDSHFOGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenylcyclohexanone undergoes Ruthenium-catalyzed reaction with tributylamine to yield 2-butyl-4-phenylcyclohexanone and 2,6-dibutyl-4-phenylcyclohexanone. Wittig reaction of 4-phenylcyclohexanone with (carbethoxymethylene)triphenylphosphorane under microwave irradiation has been investigated.

Application

4-Phenylcyclohexanone was used in the preparation of new cardo diamine monomer, 1,1-bis[4-(4-aminophenoxy)phenyl]-4-phenylcyclohexane bearing a 4-phenylcyclohexylidene unit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-Catalyzed Regioselective alpha-Alkylation of Ketones: The First Alkyl-Group Transfer from Trialkylamines to the alpha-C Atom of Ketones This work was supported by the Basic Research Program of the Korea Science and Engineering Foundation (2000-2-12200-001-3). C.S.C. gratefully acknowledges a Post-Doctoral Fellowship of Kyungpook National University (2000).
Chan Sik Cho et al.
Angewandte Chemie (International ed. in English), 40(5), 958-960 (2001-03-10)
Synthesis and Characterization of New Cardo Polyamides and Polyimides bearing a 4-Phenylcyclohexylidene Unit.
Liaw D-J, et al.
Macromolecular Chemistry and Physics, 202(6), 807-813 (2001)
Highly regioselective Wittig reactions of cyclic ketones with a stabilized phosphorus ylide under controlled microwave heating.
Wu J, et al.
Tetrahedron Letters, 45(22), 4401-4404 (2004)
Alejandro Gran-Scheuch et al.
Frontiers in microbiology, 9, 1609-1609 (2018-08-04)
Actinobacteria are an important source of commercial (bio)compounds for the biotechnological and pharmaceutical industry. They have also been successfully exploited in the search of novel biocatalysts. We set out to explore a recently identified actinomycete, Streptomyces leeuwenhoekii C34, isolated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service