Skip to Content
Merck
All Photos(3)

Key Documents

A1968

Sigma-Aldrich

Ascorbic acid 6-palmitate

meets USP testing specifications

Synonym(s):

6-O-Palmitoyl-L-ascorbic acid, L-Ascorbic acid 6-hexadecanoate, L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, NSC 402451

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O7
CAS Number:
Molecular Weight:
414.53
Beilstein:
96552
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

95.0-100.5%

form

powder

color

light yellow

application(s)

pharmaceutical (small molecule)

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

InChI

1S/C22H38O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(24)28-16-17(23)21-19(25)20(26)22(27)29-21/h17,21,23,25-26H,2-16H2,1H3/t17-,21+/m0/s1

InChI key

QAQJMLQRFWZOBN-LAUBAEHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ascorbic acid 6-palmitate/L-Ascorbyl 6-palmitate (AP) is a derivative of a synthetic lipophilic ascorbic acid.

Biochem/physiol Actions

L-Ascorbyl 6-palmitate (AP) is used as preservative in foods. In mice, AP works as a dietary antitumor promoter. 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparison of L-ascorbic acid and L-ascorbyl 6-palmitate utilization in Guinea Pigs and humans
Johnston CS, et al.
Nutrition Research (New York, N.Y.), 14(10), 1465-1471 (1994)
Charlotte Jacobsen
Annals of the New York Academy of Sciences, 1190, 141-150 (2010-04-15)
Enrichment of foods with marine omega-3 polyunsaturated fatty acids (PUFA) poses a multidisciplinary challenge to food industry and academia. Although our knowledge about possible health effects of omega-3 PUFA has increased tremendously during the last 30 years, there is still
Luciano Benedini et al.
Colloids and surfaces. B, Biointerfaces, 89, 265-270 (2011-10-15)
Ascorbyl palmitate (Asc16) in polyethyleneglycol 400 (PEG 400)-water mixtures at weight fractions (w/w) between 0.05 and 1.0 were studied by differential scanning calorimetry (DSC) and polarizing microscopy (PM) at different temperatures. The employed PEG 400-water proportions were: 0-25-50% and 75%
Gopa Mitra et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 149(4), 341-350 (2008-03-11)
The effect of supplementation of ascorbic acid through enriched zooplankton [10%, 20% and 30% ascorbyl palmitate (AP) inclusion in diet of zooplankton] on different digestive enzyme activities during ontogeny of Labeo rohita larvae was studied from 4 day to 15
Lindomar A Lerin et al.
Ultrasonics sonochemistry, 18(5), 988-996 (2011-01-29)
This work is focused on the optimization of reaction parameters for the synthesis of ascorbyl palmitate catalyzed by Candida antarctica lipase in different organic solvents under ultrasound irradiation. The sequential strategy of experimental design proved to be useful in determining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service