Skip to Content
Merck
All Photos(1)

Key Documents

A0350000

Allopurinol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1H-Pyrazolo(3,4-d)pyrimidin-4-ol, 4-Hydroxypyrazolo(3,4-d)pyrimidine, 4-Hydroxypyrazolo[3,4-d]pyrimidine, HPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

allopurinol

manufacturer/tradename

EDQM

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1NC=Nc2[nH]ncc12

InChI

1S/C5H4N4O/c10-5-3-1-8-9-4(3)6-2-7-5/h1-2H,(H2,6,7,8,9,10)

InChI key

OFCNXPDARWKPPY-UHFFFAOYSA-N

Gene Information

human ... XDH(7498)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopoeia.

Application

Allopurinol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Inhibitor of xanthine oxidase and de novo pyrimidine biosynthesis. A classical agent in treatment of hyperuricemia and gout.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Essack Mitha et al.
Rheumatology (Oxford, England), 52(7), 1285-1292 (2013-03-15)
To evaluate the efficacy and safety of IL-1 inhibitor rilonacept (IL-1 Trap) for gout flare (GF) prevention during initiation of uric acid-lowering therapy (ULT) with allopurinol in a multiregional phase 3 clinical trial. Hyperuricaemic adults (n = 248) from South
Gisele Zandman-Goddard et al.
Rheumatology (Oxford, England), 52(6), 1126-1131 (2013-02-09)
To assess the adherence and persistence with allopurinol therapy among gout patients and to identify risk factors for therapy discontinuation. The study population included adults in Maccabi Healthcare Services, a 2-million member health maintenance organization in Israel, who were diagnosed
Suborno M Ghosh et al.
Hypertension (Dallas, Tex. : 1979), 61(5), 1091-1102 (2013-04-17)
Elevation of circulating nitrite (NO2(-)) levels causes vasodilatation and lowers blood pressure in healthy volunteers. Whether these effects and the underpinning mechanisms persist in hypertension is unknown. Therefore, we investigated the consequences of systemic nitrite elevation in spontaneously hypertensive rats
Sushma Rekhraj et al.
Journal of the American College of Cardiology, 61(9), 926-932 (2013-03-02)
This study sought to ascertain if high-dose allopurinol regresses left ventricular mass (LVM) in patients with ischemic heart disease (IHD). LV hypertrophy (LVH) is common in patients with IHD including normotensive patients. Allopurinol, a xanthine oxidase inhibitor, has been shown
Sarah Leighton et al.
Experimental dermatology, 22(3), 189-194 (2013-02-08)
Exposure to solar ultraviolet (UV) radiation suppresses adaptive immune responses. This contributes to skin carcinogenesis but may protect from some autoimmune diseases. However, the molecular changes occurring within UV-exposed skin that precipitate the downstream events leading to immune suppression are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service