Skip to Content
Merck
All Photos(5)

Key Documents

I7508

Sigma-Aldrich

myo-Inositol

≥99% (GC), BioReagent

Synonym(s):

1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1907329
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

6.2 (vs air)

Quality Level

product line

BioReagent

Assay

≥99% (GC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white

mp

222-227 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

application(s)

agriculture
life science and biopharma
metabolomics
sample preparation
sample preparation
vitamins, nutraceuticals, and natural products

SMILES string

O[C@H]1[C@H](O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-

InChI key

CDAISMWEOUEBRE-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myo-Inositol, also known as i-Inositol, is a sugar alcohol and functions as a growth factor for both animals and microorganisms. Classified as a pseudovitamin, it doesn′t meet the essential vitamin criteria, as its deficiency doesn′t lead to disease conditions despite its vital role in the body. This polyol is abundantly present and serves as a structural element in eukaryotic cell secondary messengers, playing crucial roles in biological processes like insulin signal transduction and cytoskeleton transduction.

Myo-Inositol, a key component in structural lipids like phosphatidylinositol (PI) and its various phosphates, including phosphatidylinositol phosphate (PIP) lipids. It plays the role of an osmolyte, cell culture media supplement, nutrient, phosphoinositide phospholipase C inhibitor, and metabolite. It participates in cell signaling, potentially stimulating tumor cell differentiation, and is found in cell membrane phospholipids, plasma lipoproteins, and the nucleus, showcasing potential chemopreventive properties. Myo-Inositol is a versatile compound that finds application in cell culture, metabolomics and biochemical research, contributing to the exploration of cellular processes and their impact on health and disease.
myo-Inositol (MI) is a sugar-like molecule present primarily in the glial cells of the human brain. 

Application

myo-Inositol has been used:
  • in the liquid basal medium as a control for the inositol starvation of cultures
  • as a supplement in synthetic oviduct fluid medium for embryo culture
  • as a component of Fuji vitamin solution to prepare nutritive medium for the growth of sugarcane plantlets

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.
myo-Inositol (MI) serves as an osmolyte, and it aids the formation of the cell membrane and myelin sheet structures. It functions as a crucial precursor of membrane phospho-inositides and phospholipids. Elevated concentration of myo-Inositol (MI) is observed in several brain disorders such as Alzheimer′s disease, gliomatosis cerebri, and diabetes mellitus. Myo-Inositol is a constituent of membrane phospholipids, glycosylphosphatidylinositol anchors that can bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Features and Benefits

  • Ideal for Cell Biology, Metabolomics and Biochemical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert B Campenot et al.
Nature protocols, 4(12), 1869-1887 (2009-12-17)
The compartmented culture, in which primary neurons plated in a proximal compartment send their axons under silicone grease barriers and into left and right distal compartments, has enhanced the experimental capabilities of neuronal cultures. Treatments can be applied separately to
M Hallman et al.
Early human development, 10(3-4), 245-254 (1985-01-01)
According to animal studies myoinositol decreases surfactant phosphatidylglycerol and increases phosphatidylinositol. In the present study lung effluent phospholipids and serum myoinositol were analyzed in respiratory distress syndrome (RDS, 19 cases), in other lung disease (6 cases) and in 22 newborn
M Utriainen et al.
Journal of neuro-oncology, 62(3), 329-338 (2003-06-05)
The signal of choline containing compounds (Cho) in proton magnetic resonance spectroscopy (1H-MRS) is elevated in brain tumors. [11C]choline uptake as assessed using positron emission tomography (PET) has also been suggested to be higher in brain tumors than in the
Pascal M Groenen et al.
American journal of obstetrics and gynecology, 189(6), 1713-1719 (2004-01-08)
The purpose of this study was to investigate the maternal and children's myo-inositol, glucose, and zinc status in association with spina bifida risk. Sixty-three mothers and 70 children with spina bifida and 102 control mothers and 85 control children were
C Servo et al.
Diabetologia, 11(6), 575-580 (1975-12-01)
Cerebrospinal fluid (CSF) or CSF and plasma levels of sorbitol, 1,5-anhydroglucitol and myoinositol of diabetic and non-diabetic patients with normal kidney function and of diabetic and non-diabetic patients with impaired kidney function were measured by gas-liquid chromatography. The CSF sorbitol

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service