Skip to Content
Merck
All Photos(1)

Key Documents

95586

Millipore

XLD Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Xylose Lysine Deoxycholate Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
ammonium ferric citrate, 0.8 g/L
lactose, 7.5 g/L
L-lysine hydrochloride, 5 g/L
phenol red, 0.08 g/L
sodium chloride, 5 g/L
sodium deoxycholate, 2.5 g/L
sodium thiosulfate, 6.8 g/L
sucrose, 7.5 g/L
xylose, 3.5 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages
veterinary

microbiology

suitability

selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for enterobacteriaceae

Application

For the isolation and differentiation of pathogenic enterobacteriaceae in particular the species Shigella and Salmonella acc. to Taylor (1965), Taylor and Harris (1965, 1967) and Taylor and Schelhart (1967).

Preparation Note

Dissolve 56.7 g in 1 litre distilled water by heating slightly. Do NOT autoclave.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of shigellae. II. Comparison of plating media and enrichment broths.
W I Taylor et al.
American journal of clinical pathology, 44(4), 476-479 (1965-10-01)
American Journal of Clinical Pathology, 48, 350-350 (1967)
W.J. Taylor et al.
American Journal of Clinical Pathology, 48, 356-356 (1967)
Isolation of shigellae. I. Xylose lysine agars; new media for isolation of enteric pathogens.
W I Taylor
American journal of clinical pathology, 44(4), 471-475 (1965-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service