Skip to Content
Merck
All Photos(2)

Documents

1.07666

Millipore

Rappaport-Vassiliadis Salmonella Enrichment Broth (acc. harm. EP/USP/JP)

GranuCult® prime, suitable for microbiology, for Salmonella spp.

Synonym(s):

RV broth, RVS broth, Rappaport broth, Vassiliadis broth.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

sterility

non-sterile

form

granular

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

5.0-5.4 (25 °C, 42.5 g/L in H2O, after autoclaving)

bulk density

700 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Salmonella spp.

General description

Rappaport-Vassiliadis Broth, also known as RV Broth or RVS Broth, is a selective enrichment medium used in the detection and isolation of Salmonella species from a variety of samples, including food and environmental specimens. RV Broth is composed of a highly concentrated base of magnesium chloride, which inhibits many bacteria and allows for selective growth of Salmonella. The broth also contains malachite green, which further prevents the growth of non-Salmonella organisms and enhances the detection of Salmonella colonies that produce hydrogen sulfide. RV Broth is often used in combination with other media and methods for the culture, isolation, and identification of Salmonella in food and environmental samples as part of surveillance and monitoring programs for food safety.

Application

Rappaport-Vassiliadis Broth (RV Broth) is used for the detection and isolation of Salmonella species from food and environmental samples. It is particularly useful for samples that are potentially contaminated with high levels of other bacterial species, as RV Broth is designed to inhibit the growth of most bacteria while promoting the growth of Salmonella. It is a widely used method for Salmonella detection and is an important tool for food safety testing and environmental monitoring programs.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service