Skip to Content
Merck
All Photos(2)

Key Documents

94970

Supelco

Ginkgolide B

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
Beilstein:
4727611
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1C(=O)O[C@H]2[C@H](O)[C@@]34[C@H]5C[C@@H](C(C)(C)C)[C@@]36[C@@H](O)C(=O)O[C@H]6O[C@@]4(C(=O)O5)[C@@]12O

InChI

1S/C20H24O10/c1-6-12(23)28-11-9(21)18-8-5-7(16(2,3)4)17(18)10(22)13(24)29-15(17)30-20(18,14(25)27-8)19(6,11)26/h6-11,15,21-22,26H,5H2,1-4H3/t6-,7+,8-,9+,10+,11+,15+,17+,18+,19-,20-/m1/s1

InChI key

SQOJOAFXDQDRGF-MMQTXUMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginkgolide B is a diterpenoid compound belonging to the group of ginkgolides, isolated from Ginkgo biloba leaves, exhibiting a wide spectrum of pharmacological properties and is used for the treatment of Alzheimer′s disease, cerebral insufficiency and memory loss.

Application

Ginkgolide B may be used as an analytical reference standard for the quantification of the analyte in phytopharmaceutical formulations and Ginkgo biloba L. extract using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Sun Choi et al.
Metabolites, 9(7) (2019-07-10)
Ginkgo leaf extract (GLE) is a popular herbal medicine and dietary supplement for the treatment of various diseases, including cardiovascular disease. GLE contains a variety of secondary plant metabolites, such as flavonoids and terpenoids, as active components. Some of these
Leonid B Krivdin
Progress in nuclear magnetic resonance spectroscopy, 108, 17-73 (2018-12-13)
Structural applications of theoretical calculations of carbon-hydrogen spin-spin coupling constants are reviewed covering papers published mainly during the last 10-15 years with a special emphasis on the most notable studies of hybridization, substitution and stereoelectronic effects together with the investigation of
Shi-hai Xia et al.
Chinese medical journal, 120(10), 922-928 (2007-06-05)
To review the recent research progress in pharmacological actions and mechanisms of ginkgolide B. Data sources Information included in this article was identified by searching of PUBMED (1987 - 2006) online resources using the key terms "ginkgolide B", "platelet activating
Peng Lv et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 44(3), 235-240 (2011-08-23)
Cerebral ischemia and reperfusion is one of the leading causes for death and severe disabilities in the world and often lead to irreversible brain damage over later lifespan. The aim of this study was to investigate the evolution of pathological
Karyn M Maclennan et al.
Progress in neurobiology, 67(3), 235-257 (2002-08-10)
Ginkgo biloba extracts such as EGb-761 have been suggested to have a multitude of beneficial effects on CNS function, from enhancing cognitive function in dementia to facilitating recovery from acute forms of neural damage such as hypoxia/ischemia. Ginkgolide B, one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service