Skip to Content
Merck
All Photos(1)

Documents

92718

Supelco

1-(Trimethylsilyl)imidazole - Pyridine mixture

for GC derivatization, LiChropur

Synonym(s):

Silylating mixture VII, TMSI+Pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
39151701
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

description

1:4 (v/v)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

density

0.98 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

c1ccncc1.C[Si](C)(C)n2ccnc2

InChI

1S/C6H12N2Si.C5H5N/c1-9(2,3)8-5-4-7-6-8;1-2-4-6-5-3-1/h4-6H,1-3H3;1-5H

InChI key

PFVKFOIOKNIZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Trimethylsilyl)imidazole-Pyridine mixture is useful for derivatizing wet sugar samples, hindered hydroxyl groups in steroids and, in conjunction with fluorinated acylation reagents, amino acids. For moderately hindered or slowly reacting compounds, pyridine is an especially useful solvent for TMSI because of its ability to act as an HCl acceptor in silylation reactions involving organochlorosilanes.

Application

Learn more in the Product Information

Other Notes

Silylating mixture for the mild silylation of alcohols. It is often called Tri-Sil Z

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Van Ling
Journal of Chromatography A, 44, 175-175 (1969)
M A Andrews
Carbohydrate research, 194, 1-19 (1989-12-01)
Two new procedures for the gas-chromatographic analysis of monosaccharides are reported. One involves derivatization of the sugars by reaction with O-benzylhydroxylamine followed by trifluoroacetylation with N-methylbis(trifluoroacetamide) and chromatography on a DB-1701 capillary column. This technique probably provides the best resolution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service