Skip to Content
Merck
All Photos(2)

Documents

90984

Supelco

Sucralose

analytical standard

Synonym(s):

1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19Cl3O8
CAS Number:
Molecular Weight:
397.63
Beilstein:
3654410
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E955
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

InChI

1S/C12H19Cl3O8/c13-1-4-7(17)10(20)12(3-14,22-4)23-11-9(19)8(18)6(15)5(2-16)21-11/h4-11,16-20H,1-3H2/t4-,5-,6+,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

BAQAVOSOZGMPRM-QBMZZYIRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucralose is a derivative of sucrose, which is generally used as a non-nutritive sweetener in foods and beverages.
Sucralose, belonging to the class of synthetic sweeteners, is a noncarcinogenic taste enhancer, found to be 400-800 times sweeter than sucrose. It is not involved in inducing glycemic response and is virtually noncaloric. Sucralose is used as a part of a comprehensive portfolio of analytical standards to monitor natural and artificial sugar substitutes in the food and beverage industries. This product finds applications as a sweetener in a variety of food products and beverages including desserts, bakery products, canned fruits/vegetables, dressings, breakfast cereals, confectionery, medical foods, dietary supplements, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sucralose may be used as an analytical reference standard for the quantification of the analyte in:
  • Commercial sugar-free products such as Splenda and Red Raspberry Diet Rite beverage using high performance anion-exchange (HPAE) chromatography coupled with pulsed amperometric detection (PAD).
  • Water and beverage samples using liquid chromatography/time-of-flight mass spectrometry (LC/TOF-MS).

Biochem/physiol Actions

A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Food Analysis by HPLC, Second Edition (2000)
An overview of the safety of sucralose
Grotz L.V and Munro C.I
Regulatory Toxicology and Pharmacology, 55(01), 1-5 (2009)
Mitchell.AE et al.
Formulation and Production Carbonated Soft Drinks (1990)
Determination of sucralose in splenda and a sugar-free beverage using high-performance anion-exchange chromatography with pulsed amperometric detection
Hanko PV, et al.
Journal of Agricultural and Food Chemistry, 52(14), 4375-4379 (2004)
Ann-Kristin Eriksson Wiklund et al.
Chemosphere, 86(1), 50-55 (2011-10-01)
The non-calorie sweetener sucralose - sucrose containing three chlorine atoms - is intensively sweet and has become a popular substitute for sugar. Its widespread use, exceptional stability in combination with high water solubility have thus resulted in contamination of recipient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service