1.00895
Sucralose
granular, EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, NF
Pharma Manufacturing
Synonym(s):
Sucralose, 1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose
About This Item
Recommended Products
Agency
ChP
JPE
NF
Ph. Eur.
Quality Level
product line
EMPROVE® ESSENTIAL
form
fine powder
granular
particle size
≥80 μm (d90)
pH
6-8 (20 °C, 100 g/L in H2O)
mp
130 °C (decomposition)
density
1.62 g/cm3 at 20 °C
application(s)
liquid formulation
pharmaceutical
solid formulation
storage temp.
15-25°C
SMILES string
OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl
InChI
1S/C12H19Cl3O8/c13-1-4-7(17)10(20)12(3-14,22-4)23-11-9(19)8(18)6(15)5(2-16)21-11/h4-11,16-20H,1-3H2/t4-,5-,6+,7-,8+,9-,10+,11-,12+/m1/s1
InChI key
BAQAVOSOZGMPRM-QBMZZYIRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Our Sucralose is available in micronized or granular form, both pharmaceutical grade, allowing you to choose depending on your individual needs. Due to its optimal dispersion behavior, sucralose powder is typically recommended for solid dose formulations, while the fast dissolution behavior of sucralose granular makes it very well-suited for liquid formulations.
Biochem/physiol Actions
Features and Benefits
- Excellent sugar-like taste profiles
- Masking bitter aftertaste and medical off-notes with ease
- High sweetness potency offers improved cost-efficiency
- Enhanced stability
- Non-cariogenic and suitable for diabetics and patients with phenylketonuria
Legal Information
Disclaimer
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service