Skip to Content
Merck
All Photos(1)

Key Documents

90964

Supelco

Hydrogen chloride solution

3 M in methanol, for GC derivatization, LiChropur

Synonym(s):

Methanolic HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.05

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

concentration

3 M in methanol

technique(s)

gas chromatography (GC): suitable

density

0.871-0.881 g/mL at 20 °C

shipped in

dry ice

storage temp.

−20°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanolic HCl (hydrogen chloride in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.

Application

Learn more in the Product Information
Suitable for derivatization of corticosteroids, estrogens, fatty acids, hop and beer bitter acids, lipids and formic and acetic acid (in cigarette smoke).

Features and Benefits

  • Derivatization of fatty acids, particularly volatile (short chain) fatty acids
  • Clean reaction (no side reaction) with volatile by-products
  • Provides clean, fast, quantitative derivatization

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David Costalago et al.
Scientific reports, 10(1), 8517-8517 (2020-05-24)
Fish growth and survival are largely determined by the nutritional quality of their food, and the fish that grow quickly during early life stages are more likely to reproduce. To adequately estimate the quality of the prey for fish, it
Joel T Kidgell et al.
International journal of biological macromolecules, 150, 839-848 (2020-02-15)
Ulvan, a sulfated polysaccharide extracted from the green seaweed genus Ulva, has bioactive properties including an immunomodulating capacity. The immunomodulatory capacity of ulvan from Ulva ohnoi, however, has not been assessed in detail. We depolymerised purified ulvan from U. ohnoi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service