Skip to Content
Merck
All Photos(1)

Key Documents

89650

Sigma-Aldrich

p-Toluidine hydrochloride

purum, ≥99.0% (AT)

Synonym(s):

4-Methylaniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
143.61
Beilstein:
3557540
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥99.0% (AT)

form

crystals

mp

243-245 °C (lit.)

SMILES string

Cl[H].Cc1ccc(N)cc1

InChI

1S/C7H9N.ClH/c1-6-2-4-7(8)5-3-6;/h2-5H,8H2,1H3;1H

InChI key

JQKBUTDZZRGQDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Toyoda et al.
Archives of toxicology, 93(3), 753-762 (2019-01-19)
Although aromatic amines are widely used as raw materials for dyes, some of them have been concerned about carcinogenicity in the urinary bladder. We examined early changes in histopathology and the formation of γ-H2AX, a biomarker of DNA damage, in
W J Brock et al.
Toxicology letters, 54(2-3), 317-325 (1990-12-01)
The in vivo covalent binding of ortho- and para-toluidine (OT and PT) to rat hepatic macromolecules was investigated to determine if a relationship exists between the degree of binding for each isomer and its carcinogenic potency. The ortho-isomer has been
J C Kennelly et al.
Mutation research, 137(1), 39-45 (1984-07-01)
The conventional Ames assay metabolising system was confirmed to be deficient in its ability to N-acetylate. This may render the test less sensitive to compounds which normally have an acetylation step during their in vivo activation to carcinogens. The addition
Issam Ahmed Mohammed et al.
Molecules (Basel, Switzerland), 15(10), 7498-7509 (2010-10-27)
Maleic anhydride was reacted with p-aminophenol and p-toluidine in the presence of di-phosphorus pentoxide (P₂O₅) as a catalyst to produce two compounds: N-(4-hydroxy-phenyl)maleimide (I) and N-(4-methylphenyl)maleimide (II). The new azo compounds I(a-c) and II(a-c) were prepared by the reaction of
E A Kowaluk et al.
Journal of pharmaceutical sciences, 75(6), 562-570 (1986-06-01)
Aqueous solutions of several ionizable substances were stored in plastic infusion bags and the sorption of the substances monitored with time. The substances used were p-nitrophenol, p-toluidine, warfarin sodium [3-(alpha-acetonylbenzyl)-4-hydroxycoumarin sodium salt] and trifluoperazine hydrochloride (10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)phenothiazine dihydrochloride). The rate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service