Skip to Content
Merck
All Photos(1)

Key Documents

88907

Supelco

2-Methoxyethanol

analytical standard

Synonym(s):

Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

548 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.0005% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

24.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.402 (lit.)
n20/D 1.402

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxyethanol is an organic compound, which is commonly used as a solvent for industrial and consumer products.

Application

2-Methoxyethanol may be used as an analytical standard for the quantification of didanosine in biological samples using high performance liquid chromatography technique (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation of didanosine in human plasma and urine by high-performance liquid chromatography.
Knupp AC, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 533(1-2), 282-290 (1990)
Analysis of residual solvents by fast gas chromatography.
Chen KT, et al.
Journal of Chromatography A, 811(1-2), 145-150 (1998)
H-Y Chang et al.
Occupational and environmental medicine, 61(8), 697-702 (2004-07-20)
To evaluate the protective effectiveness of gloves from occupational exposure to 2-methoxyethanol (2-ME); and to examine the association of 2-methoxyacetic acid (MAA) in urine and plasma collected simultaneously from low 2-ME exposure and high 2-ME exposure workers in a semiconductor
Makoto Takei et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(2), 643-652 (2010-07-10)
Ethylene glycol monomethyl ether (EGME) is a widely used industrial solvent known to cause adverse effects to human and other mammals. Organs with high metabolism and rapid cell division, such as testes, are especially sensitive to its actions. In order
Frank Welsch
Toxicology letters, 156(1), 13-28 (2005-02-12)
Numerous experimental studies have established that only a few among the large family of ethylene glycol ethers (EGEs) elicit toxicity on reproduction in either gender. Notable are the monomethyl (EGME) and monoethyl (EGEE) ethers and their respective acetate esters whose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service