Skip to Content
Merck
All Photos(1)

Documents

87794

Sigma-Aldrich

Gram′s fuchsin solution

for microscopy

Synonym(s):

Fuchsin solution

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
4166684
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

refractive index

n20/D 1.334

density

0.999 g/mL at 20 °C

λmax

545-555 nm in 50% ethanol

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Br.CO[C@@H]1CCCN[C@H]1CC(C)=O

InChI

1S/C9H17NO2.BrH/c1-7(11)6-8-9(12-2)4-3-5-10-8;/h8-10H,3-6H2,1-2H3;1H/t8-,9+;/m0./s1

InChI key

DVSQBUUDEVNABC-OULXEKPRSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

May contain minor quantities of precipitated dye.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service