Skip to Content
Merck
All Photos(1)

Key Documents

87117

Supelco

Nervonic acid

analytical standard

Synonym(s):

cis-15-Tetracosenoic acid, Selacholeic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)13COOH (cis)
CAS Number:
Molecular Weight:
366.62
Beilstein:
1728551
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

42-43 °C (lit.)

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCCCCCC(O)=O

InChI

1S/C24H46O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h9-10H,2-8,11-23H2,1H3,(H,25,26)/b10-9-

InChI key

GWHCXVQVJPWHRF-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nervonic acids are the products of desaturation and elongation processes of various fatty acids such as palmitic acid, stearic acid, oleic acid, etc.

Application

Nervonic acid may be used as an analytical reference standard for the determination of nervonic acid in:
  • Berry seed extract samples by gas chromatography equipped with mass spectrometer detector (GC-MSD).
  • Human blood by isotope-dilution gas chromatography-negative chemical ionization-mass spectrometry (ID-GC-NCI-MS).
  • Human plasma samples employed in the clinical smoking cessation study by GC combined with time-of-flight mass spectrometry operating under the electron ionization (EI) mode.
  • Brain tissue samples by GC-MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wide-ranging alterations in the brain fatty acid complement of subjects with late Alzheimer?s disease as detected by GC-MS.
Nasaruddin ML, et al.
American Journal of Translational Research, 8(1), 154-154 (2016)
Evidence That Parietal Lobe Fatty Acids May Be More Profoundly Affected in Moderate Alzheimer?s Disease (AD) Pathology Than in Severe AD Pathology.
Nasaruddin ML, et al.
Metabolites, 8(4), 69-69 (2018)
A fully validated GC-TOF-MS method for the quantification of fatty acids revealed alterations in the metabolic profile of fatty acids after smoking cessation.
Goettel M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1041(8), 141-150 (2017)
Quantitation of trans-fatty acids in human blood via isotope dilution-gas chromatography-negative chemical ionization-mass spectrometry.
Kuiper HC, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1076(8), 35-43 (2018)
Fatty Acids Analysis Using Gas Chromatography-Mass Spectrometer Detector (GC/MSD)-Method Validation Based on Berry Seed Extract Samples.
Mazurek B, et al.
Food Analytical Methods, 10(8), 2868-2880 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service