Skip to Content
Merck
All Photos(1)

Key Documents

86838

Supelco

Tetrabutylammonium bis-trifluoromethanesulfonimidate

for electronic purposes, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[N(SO2CF3)2
CAS Number:
Molecular Weight:
522.61
Beilstein:
8247973
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for electronic purposes

Quality Level

Assay

≥99.0% (qNMR)
≥99.0%

form

crystals

mp

90-95 °C

SMILES string

FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C2F6NO4S2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-16H2,1-4H3;/q+1;-1

InChI key

CFAPFDTWIGBCQK-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application


  • The cohesive properties and pyrolysis mechanism of an aprotic ionic liquid tetrabutylammonium bis(trifluoromethanesulfonyl)imide.: This study investigates the cohesive properties and thermal decomposition mechanism of tetrabutylammonium bis(trifluoromethanesulfonyl)imide, providing insights into its stability and potential applications in analytical and materials chemistry (Liu et al., 2023).

Other Notes

Novel type of electrolyte, used e.g. in electrical double layer capacitors[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Hirshberg et al.
ACS applied materials & interfaces, 10(13), 10860-10869 (2018-03-08)
Using UV-vis spectroscopy in conjunction with various electrochemical techniques, we have developed a new effective operando methodology for investigating the oxygen reduction reactions (ORRs) and their mechanisms in nonaqueous solutions. We can follow the in situ formation and presence of
M. Ue et al.
Journal of the Electrochemical Society, 141, 3366-3366 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service