Skip to Content
Merck
All Photos(1)

Documents

00797

Sigma-Aldrich

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide

for electrochemistry, ≥99% (NMR)

Synonym(s):

Methyltri-n-octylammonium bis(trifluoromethanesulfonyl)imide, Trioctylmethylammonium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H54F6N2O4S2
CAS Number:
Molecular Weight:
648.85
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for electrochemistry

Quality Level

Assay

≥99% (NMR)

form

liquid

impurities

≤300 ppm water

refractive index

n20/D 1.439

conductivity

~0.04 mS/cm

anion traces

bromide (Br-): ≤20 mg/kg
chloride (Cl-): ≤20 mg/kg
fluoride (F-): ≤20 mg/kg
nitrate (NO3-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

InChI

1S/C25H54N.C2F6NO4S2/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-25H2,1-4H3;/q+1;-1

InChI key

LAGQNGWYNLUQRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide is an ionic liquid used as an electrolyte in various electrochemical applications including sensors, batteries, and capacitors due to its diverse properties such as Low volatility, high conductivity, excellent electrochemical window and ability to solvate multiple species.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service