Skip to Content
Merck
All Photos(1)

Key Documents

86490

Supelco

2,2′:6′,2′′-Terpyridine

for spectrophotometric det. of Ag, Fe, Ru, ≥98.5%

Synonym(s):

α,α′,α″-Tripyridyl, 2,6-Di(2-pyridyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3
CAS Number:
Molecular Weight:
233.27
Beilstein:
11199
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.5% (GC)
≥98.5%

form

crystals

quality

for spectrophotometric det. of Ag, Fe, Ru

technique(s)

UV/Vis spectroscopy: suitable

mp

89-91 °C (lit.)
90-93 °C

solubility

dioxane: 0.1 g/mL, clear

cation traces

Ag: ≤5 mg/kg
Ba: ≤5 mg/kg
Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

c1ccc(nc1)-c2cccc(n2)-c3ccccn3

InChI

1S/C15H11N3/c1-3-10-16-12(6-1)14-8-5-9-15(18-14)13-7-2-4-11-17-13/h1-11H

InChI key

DRGAZIDRYFYHIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent for the spectrophotometric det. of Ag and Ru

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.C. Kamra et al.
Analytica Chimica Acta, 81, 177-177 (1976)
E. Gagliardi et al.
Mikrochimica Acta, 1175-1175 (1964)
S Rahuel-Clermont et al.
Biochemistry, 36(19), 5837-5845 (1997-05-13)
The insulin hexamer is an allosteric protein capable of undergoing transitions between three conformational states: T6, T3R3, and R6. These transitions are mediated by the binding of phenolic compounds to the R-state subunits, which provide positive homotropic effects, and by
Teng-Yuan Dong et al.
Langmuir : the ACS journal of surfaces and colloids, 20(21), 9340-9347 (2004-10-06)
Spectroscopic and electrochemical characterizations of ferrocene- and biferrocene-functionalized terpyridine octanethiolate monolayer-protected clusters were investigated and reported. The electrochemical measurements of Ru2+ coordinated with 4'-ferrocenyl-2,2':6',2' '-terpyridine and 4'-biferrocenyl-2,2':6',2' '-terpyridine complexes were dominated by the Ru2+/Ru3+ redox couple (E(1/2) at approximately 1.3
Wah-Leung Tong et al.
Dalton transactions (Cambridge, England : 2003), (24)(24), 4741-4746 (2009-06-11)
A modular design approach has been utilized to develop molecular 'pockets' featuring three integrated components, namely a phosphorescent Pt(II)-pi(organic) moiety, a suitable receptor group, and a rigid, conjugated connecting backbone, and the cavities therein are examined by NMR spectroscopy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service