Skip to Content
Merck
All Photos(1)

Key Documents

496375

Sigma-Aldrich

4′-(4-Methylphenyl)-2,2′:6′,2′′-terpyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3
CAS Number:
Molecular Weight:
323.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

mp

167-170 °C (lit.)

SMILES string

Cc1ccc(cc1)-c2cc(nc(c2)-c3ccccn3)-c4ccccn4

InChI

1S/C22H17N3/c1-16-8-10-17(11-9-16)18-14-21(19-6-2-4-12-23-19)25-22(15-18)20-7-3-5-13-24-20/h2-15H,1H3

InChI key

IDJYBUVHZHLIIT-UHFFFAOYSA-N

General description

4′-(4-Methylphenyl)-2,2′:6′,2′′-terpyridine is a tridentate oligopyridine ligand.[1] Its crystalline structure has been investigated.[2]

Application

4′-(4-Methylphenyl)-2,2′:6′,2′′-terpyridine may be used as a ligand in the synthesis of:
  • terpyridine Ru(II) complexes[3]
  • terpyridine based Cd(II) and Hg(II) complexes[1]
  • heterobimetallic Ru(II)-Os(II) complexes[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yamamoto et al.
Life sciences, 59(25-26), 2139-2146 (1996-01-01)
We have identified 4'-(4-methylphenyl)-2,2':6',2-terpyridine: trihydrochloride (SS701), which belongs to a family of a small unique neuroprotective agents. SS701 accelerated the production of nerve growth factor (NGF) in cultured astroglial cells, dose- and time-dependently. In in vivo studies, SS701, when administered
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine.
Liu HG, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(12), o4876-o4876 (2007)
Photoinduced intramolecular energy transfer and anion sensing studies of isomeric RuII OsII complexes derived from an asymmetric phenanthroline-terpyridine bridge.
Maity D, et al.
Dalton Transactions, 43(4), 1829-1845 (2014)
Li Jiao Liang et al.
The Analyst, 137(4), 953-958 (2011-12-21)
Pyrophosphate ion (PPi) is crucial in varieties of biological processes and industrial applications, and thus it is very important how to recognize it with high selectivity. In this contribution, one terpyridine (tpy)-based fluorescent molecule, 4-(methylphenyl)-2,2':6',2''-terpyridine (mptpy), has been reported to
Synthesis, crystal structure and thermal properties of Cd(II) and Hg(II) terpyridine based compounds.
Saghatforoush L, et al.
Main Group Chemistry, 13(1), 29-39 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service