81123
Poly(acrylic acid sodium salt)
analytical standard, for GPC, 16,000
Synonym(s):
Poly(sodium acrylate), Sodium polyacrylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
for GPC
Quality Level
shelf life
limited shelf life, expiry date on the label
mol wt
Mn ~12800
Mp ~16000
Mw ~18100
technique(s)
gel permeation chromatography (GPC): suitable
Mw/Mn
~1.41
format
neat
storage temp.
2-8°C
SMILES string
[Na+].[O-]C(=O)C=C
InChI
1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1
InChI key
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lei Liu et al.
ACS nano, 7(2), 1368-1378 (2013-01-04)
Hierarchical FeOOH nanostructure array films constructed by different nanosized building blocks can be synthesized at the air-water interface via a bio-inspired gas-liquid diffusion method. In this approach, poly(acrylic acid) (PAA) as a crystal growth modifier plays a crucial role in
Daniel L Miller et al.
The Annals of thoracic surgery, 95(3), 1050-1056 (2013-01-22)
Skeletal chest wall reconstruction can be a challenge, depending on the indication, location, and health of the patient; various materials are available. Recently, biomaterials that are remodelable (bovine pericardium patch; Veritas, Synovis Life Technologies Inc, St Paul, MN) or absorbable
Kang Zhong et al.
Carbohydrate polymers, 92(2), 1367-1376 (2013-02-13)
Phosphate rock (PHR), a traditional fertilizer, is abundant, but is hard to be utilized by plants. To improve the utilization of PHR, and to integrate water-retaining and controlled-release fertilizers, an agricultural superabsorbent polymer based on sulfonated corn starch/poly (acrylic acid)
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Imran Khimji et al.
Chemical communications (Cambridge, England), 49(13), 1306-1308 (2013-01-11)
The melting temperature of duplex DNA is much higher in polyanions than in non-ionic polymers with similar ionic strength, suggesting an additional electrostatic contribution on top of the excluded volume effect.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service