Skip to Content
Merck
All Photos(1)

Documents

416010

Sigma-Aldrich

Poly(acrylic acid, sodium salt) solution

average Mw ~1,200, 45 wt. % in H2O

Synonym(s):

PAA, Sodium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CO2Na)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mw ~1,200

concentration

45 wt. % in H2O

refractive index

n20/D 1.43

density

1.32 g/mL at 25 °C

SMILES string

[Na]OC(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Poly(acrylic acid, sodium salt) solution has been used to study the mechanism of formation of supramolecularaggregates of polyacrylic acid(PAA) in the presence of Fe(lll). This supramolecular association traps all Fe(lll) and prevents the precipitation of Fe (OH)3.The chelation capacity of PAA can be exploited to inhibit the precipitation ofhematite and other iron oxides.

Itcan be used as a draw solute in forward osmosis process due to its flexibilityin structural configuration and high solubility in water. They can be preferredover conventional ionic salts as they can produce comparable water flux andmuch lower reverse leakages.

It can also be used to study the effect of the molecular weight of PAAon crystal nucleation and the growth of CaCO3 during thecrystallization process.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Liu et al.
ACS nano, 7(2), 1368-1378 (2013-01-04)
Hierarchical FeOOH nanostructure array films constructed by different nanosized building blocks can be synthesized at the air-water interface via a bio-inspired gas-liquid diffusion method. In this approach, poly(acrylic acid) (PAA) as a crystal growth modifier plays a crucial role in
Michael Dietzsch et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 3080-3088 (2013-02-08)
Scale formation, the deposition of certain minerals such as CaCO3, MgCO3, and CaSO4·2H2O in industrial facilities and household devices, leads to reduced efficiency or severe damage. Therefore, incrustation is a major problem in everyday life. In recent years, double hydrophilic
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Serhat Elcicek et al.
Experimental parasitology, 133(3), 237-242 (2012-12-15)
Polyacrylic acid (PAA) is one of the anionic synthetic polyelectrolytes and is used in various immunological and pharmaceutical applications. PAA has been used as adjuvant in veterinary vaccines, in particular. However, to our knowledge, there are no studies that document
Priscila Stona et al.
The journal of adhesive dentistry, 15(3), 221-227 (2013-04-06)
To examine the influence of 11.5% polyacrylic acid pretreatment on the interface and bond strength of self-adhesive resin cements (Maxcem Elite, RelyX Unicem, SeT) to dentin. Fifty-six third molars were randomly divided into seven groups: RelyX ARC as control (ARC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service