Skip to Content
Merck
All Photos(1)

Key Documents

78780

Sigma-Aldrich

Phenyl isothiocyanate

for HPLC derivatization, the detection of alcohols and amines, ≥99.0%

Synonym(s):

PITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NCS
CAS Number:
Molecular Weight:
135.19
Beilstein:
471392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0%

form

liquid

quality

for HPLC derivatization, the detection of alcohols and amines

refractive index

n20/D 1.649
n20/D 1.6515 (lit.)

bp

218 °C (lit.)

mp

−21 °C (lit.)

solubility

water: insoluble

density

1.13 g/mL at 20 °C
1.132 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=Nc1ccccc1

InChI

1S/C7H5NS/c9-6-8-7-4-2-1-3-5-7/h1-5H

InChI key

QKFJKGMPGYROCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenyl isothiocyanate is an aromatic isothiocyanate. It participates in dehydration reactions of alcohols. It is widely used for synthesis of various biologically important heterocyclic compounds.

Application

Phenyl isothiocyanate may be employed as a derivatization reagent for high-performance liquid chromatographic (HPLC) analysis of various amphetamine derivatives in body fluids.
Derivatizing reagent for primary and secondary amines. Used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC.

Caution

Store under Argon

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amin Zolali et al.
Combinatorial chemistry & high throughput screening, 17(7), 610-613 (2014-03-19)
An efficient, one-pot and three-component synthesis of biologically important heterocyclic compounds is described from the reaction of primary amines and phenyl isothiocyanate in the presence of acryloyl chloride at room temperature without the need to use any catalyst.
One-pot dehydrations using phenyl isothiocyanate.
Majetich G, et al.
Tetrahedron Letters, 56(23), 3326-3329 (2015)
Determination of phenylisothiocyanate derivatives of amphetamine and its analogues in biological fluids by HPLC-APCI-MS or DAD.
Bogusz MJ, et al.
Journal of Analytical Toxicology, 21(1), 59-69 (1997)
Tomoyuki Oe et al.
Rapid communications in mass spectrometry : RCM, 24(2), 173-179 (2009-12-17)
This manuscript describes the results of a preliminary experiment performed as 'proof of concept' of a novel approach to absolute quantitation of proteins without the use of standard proteins. Absolute quantitation remains a challenging issue in the proteomics field. Therefore
Jiapeng Leng et al.
Journal of the American Society for Mass Spectrometry, 22(7), 1204-1213 (2011-09-29)
A multifunctional isothiocyanate-based isotope labeling reagent, [d (0)]-/[d (6)]-4,6-dimethoxy pyrimidine-2-isothiocyanate (DMPITC), has been developed for accurate N-terminus identification in peptide sequencing and comparative protein analysis by ESI Ion-trap TOF mass spectrometry. In contrast with the conventional labeling reagent phenyl isothiocyanate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service