Skip to Content
Merck
All Photos(1)

Key Documents

78013

Supelco

4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone

analytical standard

Synonym(s):

4-(Methylnitrosamino)-1-(3-pyridinyl)-1-butanone, NNK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O2
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CN(CCCC(=O)c1cccnc1)N=O

InChI

1S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8H,3,5,7H2,1H3

InChI key

FLAQQSHRLBFIEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aymeric Monteillier et al.
Scientific reports, 8(1), 14257-14257 (2018-09-27)
Lung cancer is the most lethal cancer in the world. About 80% of lung cancer deaths are linked to tobacco use. As a complement to tobacco control, efficient chemoprevention strategies are needed to tackle lung cancer epidemic. Resveratrol is one
Harald Hundsberger et al.
BMC genomics, 18(1), 450-450 (2017-06-10)
Recombinant peptide chips could constitute a versatile complementation to state-of-the-art in situ (chemical on-chip) synthesis, particle-based printing, or pre-manufactured peptide spotting. Bottlenecks still impeding a routine implementation - from restricted peptide lengths, low diversity and low array densities to high
Matt S Melvin et al.
ACS omega, 5(28), 17565-17575 (2020-07-28)
Exposure to diacetyl (DA) has been linked to the respiratory condition bronchiolitis obliterans. Previous research has demonstrated that DA and other α-dicarbonyl compounds can be detected in both the e-liquids and aerosols of e-vapor products (EVPs). While some EVP manufacturers
Thomas Thisted et al.
BMC biotechnology, 19(1), 56-56 (2019-08-04)
Smoking and tobacco use continue to be the largest preventable causes of death globally. A novel therapeutic approach has recently been proposed: administration of an enzyme that degrades nicotine, the main addictive component of tobacco, minimizing brain exposure and reducing
Huawei Qiu et al.
mAbs, 11(7), 1266-1275 (2019-06-15)
Deamidation evaluation and mitigation is an important aspect of therapeutic antibody developability assessment. We investigated the structure and function of the Asn-Gly deamidation in a human anti-CD52 IgG1 antibody light chain complementarity-determining region 1, and risk mitigation through protein engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service