Skip to Content
Merck
All Photos(1)

Key Documents

175870

Sigma-Aldrich

Benzene-D6

"100%", 99.96 atom % D

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

isotopic purity

99.96 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene-d6 (C6D6) is a deuterated derivative of benzene. It is 100% isotopically enriched NMR (Nuclear Magnetic Resonance) solvent. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity. Its Soret coefficient S(T) has been measured by the transient holographic grating technique. Its first resonance Raman spectrum and infrared absorption intensities (in the range of 5000-450cm-1) have been reported. It participates in the synthesis of isotopically labeled samples.

Application

Benzene-d6 may be used as a 1H and 13C-NMR solvent. It may also be used for the synthesis of isotopically labeled samples and for the internal calibration of benzene stock solution during headspace-solid-phase microextraction.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Structure-property relationships in nematic gold nanoparticles.
Cseh L and Mehl GH.
Journal of Materials Chemistry, 17(4), 311-315 (2007)
Resonance Raman scattering of benzene and benzene-d6 with 212.8 nm excitation.
Ziegler LD and Hudson B.
J. Chem. Phys. , 74(2), 982-992 (1981)
Surprising Titanium complexes bearing η2-pyrazolato ligands: synthesis, structure, and molecular orbital studies.
Guzei IA, et al.
Journal of the American Chemical Society, 119(14), 3387-3388 (1997)
Biomonitoring of benzene and toluene in human blood by headspace-solid-phase microextraction.
Schimming E, et al.
Fresenius Journal of Analytical Chemistry, 363(1), 88-91 (1999)
G Wittko et al.
The Journal of chemical physics, 123(1), 014506-014506 (2005-07-23)
The Soret coefficients S(T) of the liquids acetone, benzene, benzene-d1, 1,3,5-benzene-d3, benzene-d5, benzene-13C6, benzene-d6, n-hexane, toluene, 1,2,3,4-tetrahydronaphthalene, isobutylbenzene, and 1,6-dibromohexane have been measured in protonated and perdeuterated cyclohexane by a transient holographic grating technique. It has been found that S(T)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service