Skip to Content
Merck
All Photos(1)

Key Documents

77367

Sigma-Aldrich

ElectroGreen® (−)-Ethyl L-lactate

photoresist grade, ≥99.0%

Synonym(s):

(−)-Ethyl (S)-2-hydroxypropionate, (S)-(−)-2-Hydroxypropionic acid ethyl ester, L-Lactic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1720839
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

product name

(−)-Ethyl L-lactate, photoresist grade, ≥99.0%

grade

photoresist grade

Quality Level

vapor density

4.1 (vs air)

vapor pressure

2 mmHg ( 20 °C)
5 mmHg ( 30 °C)

Assay

≥99.0% (Sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D −10.5±1°, neat

autoignition temp.

752 °F

expl. lim.

1.5 %, 100 °F

impurities

≤0.1% water

evapn. residue

<0.01%

refractive index

n20/D 1.4130 (lit.)
n20/D 1.413

bp

154 °C (lit.)

mp

−26 °C (lit.)

density

1.034 g/mL at 20 °C (lit.)

cation traces

Ca: ≤0.01 mg/kg
Cr: ≤0.01 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.01 mg/kg
K: ≤0.01 mg/kg
Mg: ≤0.01 mg/kg
Mn: ≤0.01 mg/kg
Na: ≤0.03 mg/kg
Ni: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg

SMILES string

C[C@H](O)C(OCC)=O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

biodegradable solvent replacement for optical and microelectronical purposes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiahua Li et al.
Journal of hazardous materials, 181(1-3), 673-678 (2010-06-18)
In order to explore a practical approach to the remediation of the cadmium (Cd)-contaminated soil in the lower reaches of the Yangtze River, we evaluated the effects of a local willow (Salix x aureo-pendula CL 'J1011') of absorbing, accumulating, and
Chiew Lin Yap et al.
Journal of environmental sciences (China), 24(6), 1064-1075 (2012-01-01)
Solubility data of recalcitrant contaminants in cosolvents is essential to determine their potential applications in enhanced soil remediation. The solubilities of phenanthrene, anthracene, fluoranthene and benzo[a]pyrene in ethyl lactate/water and ethanol/water mixtures were measured using equilibrium techniques. The cosolvency powers
Hongyan Guo et al.
Journal of hazardous materials, 174(1-3), 59-63 (2009-09-29)
A new approach using aqueous ethyl lactate-modified [S,S]-ethylenediaminedisuccinic acid (EDDS) washing solutions for the removal of Cu from field-contaminated soils was examined in the laboratory. At pH 7 the maximum extraction efficiencies of Cu were about 41% by EDDS solution
Betty K Ishida et al.
Journal of agricultural and food chemistry, 57(3), 1051-1059 (2009-01-14)
Few environmentally friendly solvents are available to extract carotenoids for use in foods. The most effective known solvents are products of the petroleum industry and toxic for human consumption. Yet carotenoid extracts are desirable for use in dietary supplements and
Marc R Elie et al.
Journal of hazardous materials, 248-249, 150-158 (2013-01-29)
The present study evaluates the capability of an activated-magnesium metal and protic co-solvents to promote the reductive degradation of three different polycyclic aromatic hydrocarbons, specifically pyrene, benzo[k]fluoranthene and benzo[g,h,i]perylene. Multivariate analyses demonstrated that the kinetics of degradation was affected by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service