Skip to Content
Merck
All Photos(5)

Key Documents

1.09639

Supelco

Ethyl(-)-L-lactate

greener alternative

EMPLURA®

Synonym(s):

Ethyl(-)-L-lactate, L(-)-Lactic acid ethyl ester, (S)-(-)-2-Hydroxypropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-694-1
NACRES:
NA.05

vapor pressure

1.6 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

form

liquid

autoignition temp.

400 °C

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

1.5-16.4 % (v/v)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

4 (20 °C, 50 g/L in H2O)

kinematic viscosity

2.7 cSt(25 °C)

bp

154 °C/1013 hPa

mp

-25 °C

transition temp

flash point 53 °C

density

1.03 g/cm3 at 20 °C

greener alternative category

storage temp.

2-25°C

SMILES string

O[C@@H](C)C(=O)OCC

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

General description

EMPLURA®
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a safer and more sustainable alternative to ethyl acetate and acetone. It is an ester of natural L-lactic acid, which is produced by fermentation of sugar and thus aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information.

Features and Benefits

Greener solvent

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service