Skip to Content
Merck
All Photos(2)

Key Documents

70173

Millipore

Casein Peptone suitable for biotechnology and microbiology

Pancreatic digest of casein from bovine milk, ≥12% Nitrogen

Synonym(s):

Peptone from animal tissue, Peptone from casein and other animal proteins

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040102
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

Peptone from casein and other animal proteins, suitable for microbiology

biological source

bovine milk

Quality Level

form

powder

composition

total nitrogen (N), ≥12%

ign. residue

≤9%

loss

≤10% loss on drying

pH

6.5-7.5 (2% in H2O)

solubility

H2O: 2%, clear, yellow (and faint yellow and faint brown)

application(s)

food and beverages
microbiology

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rüdiger W Maschke et al.
Bioengineering (Basel, Switzerland), 9(8) (2022-07-28)
Shake flasks remain one of the most widely used cultivation systems in biotechnology, especially for process development (cell line and parameter screening). This can be justified by their ease of use as well as their low investment and running costs.
Polysaccharide metabolism regulates structural colour in bacterial colonies.
van de Kerkhof, et al.
Journal of the Royal Society, Interface / the Royal Society, 19, 20220181-20220181 (2022)
Mesut Taskin et al.
International journal of food sciences and nutrition, 63(5), 597-602 (2011-12-06)
This study was performed to investigate the usability of chicken feather hydrolysate (Chicken feather peptone (CFP)) as substrate for mycelial biomass and extracellular polysaccharides (EPS) production from edible mushroom Morchella esculenta. The ability of CFP to support biomass and EPS
M Taskin et al.
Journal of applied microbiology, 111(4), 826-834 (2011-07-19)
Peptones are one of the most expensive constituents of microbial media. This study was undertaken to prepare the peptone from waste chicken feathers through a new process. The chemical analysis of chicken feather peptone (CFP) was performed. The ability of
Saoussen Turki et al.
Bioresource technology, 100(10), 2724-2731 (2009-02-04)
In this work the effect of several organic nitrogen sources on lipase production in Yarrowia lipolytica LgX64.81 overproducing mutant was studied. Among them, tryptone and peptone showed the most prominent stimulatory effect. Interestingly, only tryptic and peptic casein digest were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service