Skip to Content
Merck
All Photos(1)

Key Documents

1.02239

Millipore

Casein Peptone for Biotechnology and Microbiology

Pancreatically Digested, Prime, for Biotechnology and Microbiology, 13.8 - 15.8% Total N

Synonym(s):

Peptone from casein pancreatically digested, Fermtech base materials for biotechnology

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
293-428-4
NACRES:
NA.74

Pricing and availability is not currently available.

Product Name

Peptone from casein pancreatically digested, free from sulfonamide antagonists, suitable for microbiology

biological source

bovine milk

Quality Level

form

solid

solubility

800 g/L

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

15-25°C

Analysis Note

Appearance: Lightly yellow to lightly brownishyellow fine powder. Characteristic, not rotten smell.
pH-value (5 %; water): 5.0 - 6.0
Total nitrogen (N) (Kjeldahl): 13.8 - 15.8 %
Amino nitrogen (as N): 4.7 - 6.7 %
Sulfated ash (800 °C): ≤ 5.0 %
Loss on drying (105 °C): ≤ 6.5 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service