Skip to Content
Merck
All Photos(2)

Key Documents

A96150

Sigma-Aldrich

Azelaic acid

technical grade, 80%

Synonym(s):

1,7-Heptanedicarboxylic acid, Anchoic acid, Nonanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)7CO2H
CAS Number:
Molecular Weight:
188.22
Beilstein:
1101094
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

6.5 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

80%

form

powder

bp

286 °C/100 mmHg (lit.)

mp

109-111 °C (lit.)

SMILES string

OC(=O)CCCCCCCC(O)=O

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)

InChI key

BDJRBEYXGGNYIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azelaic acid is used as a therapeutic agent in dermatology.

Application

Azelaic acid, a bolaamphiphile, is generally used as a spacer/linker in synthetic chemistry. Some of the applications include:
  • Synthesis of pH-responsive metallo-hydrogels as potential dye-adsorbing agents, and vitamin B12 carriers.
  • Synthesis of fluorescent uranyl (UO22+) coordination polymers.
  • Synthesis of various metal-organic frameworks (MOFs) and supramolecular macrocyclic adducts.
  • Conversion of hydrophobic up-converting rare-earth nanophosphors (UCNPs) into water-soluble, carboxylic acid-functionalized analogs to facilitate conjugation with biomolecules such as streptavidin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluation of the antibacterial efficacy of diesters of azelaic acid.
Charnock C, et al.
European Journal of Pharmaceutical Sciences, 21(5), 589-596 (2004)
Crystal engineering with the uranyl cation I. Aliphatic carboxylate coordination polymers: Synthesis, crystal structures, and fluorescent properties.
Borkowski LA and Cahill CL
Crystal Growth & Design, 6(10), 2241-2247 (2006)
Solid-state preparation of hybrid organometallic-organic macrocyclic adducts with long chain dicarboxylic acids.
Lucaa G, et al.
Chemical Communications (Cambridge, England), 8(37), 3877-3879 (2006)
Pillaring Zn-Triazolate Layers with Flexible Aliphatic Dicarboxylates into Three-Dimensional Metal- Organic Frameworks.
Lin Y-Y, et al.
Crystal Growth & Design, 8(10), 3673-3679 (2008)
pH-responsive, bolaamphiphile-based smart metallo-hydrogels as potential dye-adsorbing agents, water purifier, and vitamin B12 carrier.
Ray S, et al.
Chemistry of Materials, 19(7), 1633-1639 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service