Skip to Content
Merck
All Photos(1)

Documents

65996

Supelco

m-Cresol

analytical standard

Synonym(s):

1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506719
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.72 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

1036 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.06-1.35 %, 150 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)
n20/D 1.542

bp

203 °C (lit.)

mp

8-10 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1cccc(O)c1

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here
m-Cresol is an isomer form of Cresol and is mainly used as an intermediate in the manufacture of chemicals. It is toxic in nature.

Application

It was used as reference standard solution for determination of phenolic compounds in tyrosinase biosensor using amperometric detection under self-assembled monolayers-based bioelectrode flow-injection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Amperometric flow-injection determination of phenolic compounds at self-assembled monolayer-based tyrosinase biosensors.
Campuzano, Susana, et al.
Analytica Chimica Acta, 494.1, 187-197 (2003)
Christian Dye
Bronopol, Resorcinol, m-Cresol and Triclosan in the Nordic Environment, 20-20 (2007)
P Saravanan et al.
Bioresource technology, 99(18), 8553-8558 (2008-05-13)
An internal loop airlift reactor (ILALR) is developed and studied for biodegradation of phenol/m-cresol as single and dual substrate systems under batch and fed batch operation using an indigenous mixed microbial strain, predominantly Pseudomonas sp. The results showed that the
Chiu-Po Chan et al.
Toxicology, 208(1), 95-104 (2005-01-25)
Cresol is a well-known environmental pollutant, toluene metabolite, uremic toxicant and accidental poisoning product. Formocresol, a preparation of formalin and cresol, is also used as a root canal medicament and for pulpotomy of primary teeth. However, little is known about
Laurence A Angel et al.
The journal of physical chemistry. A, 110(35), 10392-10403 (2006-09-01)
Energy-resolved, competitive threshold collision-induced dissociation (TCID) methods are used to measure the gas-phase acidities of phenol, 3-methylphenol, 2,4,6-trimethylphenol, and ethanoic acid relative to hydrogen cyanide, hydrogen sulfide, and the hydroperoxyl radical using guided ion beam tandem mass spectrometry. The gas-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service