Skip to Content
Merck
All Photos(1)

Key Documents

65760

Sigma-Aldrich

N-Methylaniline

purum, ≥98.0% (GC)

Synonym(s):

Monomethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH3
CAS Number:
Molecular Weight:
107.15
Beilstein:
741982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

refractive index

n20/D 1.571 (lit.)
n20/D 1.571

bp

196 °C (lit.)

mp

−57 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNc1ccccc1

InChI

1S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8H,1H3

InChI key

AFBPFSWMIHJQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Liver,spleen,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Itoh et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 40(5), 1013-1019 (2005-05-13)
Photolysis of 1,3,5-trichlorobenzene (TCB) in moderate concentration (0.5 mM) in the presence of several additives was examined in 10 mM of cationic and nonionic surfactant solutions. Additions of small amounts of hydrophobic additives, n-dodecanethiol (1 mM), n-dodecyldimethylamine (<2.5 mM), and
Cai Jianguo et al.
Chemosphere, 61(4), 502-509 (2005-10-06)
In the present study, a hydrophilic bifunctional polymeric resin (LS-2) with sulfonic groups was synthesized, and the adsorption performance of three aniline compounds, aniline, 4-methylaniline, and 4-nitroaniline onto LS-2 was compared with that on the commercial Amberlite XAD-4. The uptake
Cai Jianguo et al.
Journal of hazardous materials, 124(1-3), 173-180 (2005-06-02)
In this paper a new bifunctional polymeric resin (LS-2) was synthesized by introducing sulfonic groups onto the surface of the resin during the post-crossing of chloromethyl low crosslinking macroporous poly-styrene resin, and the comparison of the adsorption properties of LS-2
Yang-fang Ye et al.
Journal of environmental sciences (China), 16(2), 343-347 (2004-05-13)
A bacterium (designated strain Y1) degrading acetanilide herbicide mefenacet was isolated from aerobic sludge. Based on the analyses of partial 16S rRNA gene, cellular fatty acid and BIOLOG-GN, and general physiological and biochemical characteristics, strain Y1 was identified as Sphingobacterium
Allison G Condie et al.
ChemMedChem, 7(12), 2194-2203 (2012-11-09)
Myelination is a fundamental biological process in the vertebrate nervous system. Damage to or malformation of myelin can lead to various neurological diseases; for example, demyelination in the spinal cord is a major cause of paralysis of patients suffering from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service