Skip to Content
Merck
All Photos(1)

Key Documents

56563

Supelco

tert-Butyldimethylsilyl trifluoromethanesulfonate

for GC derivatization, LiChropur, ≥98.0% (T)

Synonym(s):

TBDMS triflate, Trifluoromethanesulfonic acid tert-butyldimethylsilylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
264.34
Beilstein:
2370068
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥98.0% (T)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.385 (lit.)
n20/D 1.385

bp

65-67 °C/12 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

InChI

1S/C7H15F3O3SSi/c1-6(2,3)15(4,5)13-14(11,12)7(8,9)10/h1-5H3

InChI key

WLLIXJBWWFGEHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service