Skip to Content
Merck
All Photos(1)

Key Documents

06735

Supelco

tert-Butyldimethylsilyl chloride

for GC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

tert-Butyl(chloro)dimethylsilane, tert-Butyldimethylchlorosilane, TBDMSCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Beilstein:
505999
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥99.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

bp

125 °C (lit.)

mp

86-89 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)Cl

InChI

1S/C6H15ClSi/c1-6(2,3)8(4,5)7/h1-5H3

InChI key

BCNZYOJHNLTNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1A

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Wakimoto et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2898-2909 (2014-04-10)
Isocitrate dehydrogenase (IDH) gene mutations occur in low-grade and high-grade gliomas. We sought to identify the genetic basis of malignant phenotype heterogeneity in IDH-mutant gliomas. We prospectively implanted tumor specimens from 20 consecutive IDH1-mutant glioma resections into mouse brains and
Some observations on the t-butyldimethylchlorosilane derivatization reaction.
K Bricknell et al.
Biochemical medicine, 23(1), 119-121 (1980-02-01)
P Huttenloch et al.
Environmental science & technology, 35(21), 4260-4264 (2001-11-23)
The efficacy of the surface modification of natural diatomite and zeolite material by chlorosilanes is demonstrated. Chlorosilanes used were trimethylchlorosilane (TMSCI), tert-butyldimethylchlorosilane (TBDMSCI), dimethyloctadecylchlorosilane (DMODSCI), and diphenyldichlorosilane (DPDSCI) possessing different headgroups and chemical properties. Silanol groups of the diatomite and
C Tellis et al.
Journal of biochemistry, 119(4), 823-827 (1996-04-01)
1-O-Hexadecyl-2-acetyl-sn-glycerol, the immediate precursor of platelet- activating factor (PAF) in its de novo formation, was detected in the protozoon Tetrahymena pyriformis. It was purified from the total lipid extract by TLC, after successive developments in two different solvent systems. Characterization
Eva C Hartmann et al.
Archives of toxicology, 85(6), 601-606 (2010-10-19)
A dose of 0.99 mg d(3)-acrylamide (d(3)-AA) (13.2 μg/kg body weight) was ingested by a healthy male volunteer. Urine samples were collected over a period of 46 h after the intake and analyzed for the hydrolysis product of glycidamide (GA), 2,3-dihydroxy-propionamide (OH-PA), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service