Recommended Products
grade
analytical standard
Quality Level
vapor density
3 (vs air)
Assay
≥98.0% (GC)
autoignition temp.
725 °F
shelf life
limited shelf life, expiry date on the label
expl. lim.
10 %
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.410±0.002 (lit.)
n20/D 1.411
bp
130 °C mmHg (lit.)
density
0.819 g/mL at 20 °C (lit.)
0.815 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCC(C)CO
InChI
1S/C5H12O/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3
InChI key
QPRQEDXDYOZYLA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
108.5 °F - closed cup
Flash Point(C)
42.5 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service