Skip to Content
Merck
All Photos(1)

Key Documents

04623

Supelco

Furfural

analytical standard

Synonym(s):

2-Furaldehyde, Furan-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
Beilstein:
105755
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

vapor density

3.31 (vs air)

vapor pressure

13.5 mmHg ( 55 °C)

Assay

≥98.5% (GC)

autoignition temp.

599 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

19.3 %

impurities

≤0.5% water

refractive index

n20/D 1.522-1.528
n20/D 1.525 (lit.)

bp

162 °C (lit.)

mp

−36 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccco1

InChI

1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H

InChI key

HYBBIBNJHNGZAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furfural is a furan derivative obtained from renewable biomass-derived carbohydrates.[1]

Application

Furfural is a sustainable alternative for petroleum-based building blocks used in the production of fine chemicals, plastics, etc[1] and also offers a promising, rich platform for lignocellulosic biofuels.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Tallarico et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 250, 119367-119367 (2021-01-06)
Chemocatalytic conversion of cellulose into lactic acid is a valuable alternative to simple sugar fermentation. Nevertheless, the procedures still need optimization to be translated to the industrial scale. Such translation would benefit by on-line monitoring of reaction parameters by fast
Production of 5-hydroxymethylfurfural and furfural by dehydration of biomass-derived mono-and poly-saccharides.
Chheda JN, et al.
Green Chemistry, 9(4), 342-350 (2007)
Reetta Karinen et al.
ChemSusChem, 4(8), 1002-1016 (2011-07-06)
Furfurals are important intermediates in the chemical industry. They are typically produced by homogeneous catalysis in aqueous solutions. However, heterogeneously catalyzed processes would be beneficial in view of the principles of green chemistry: the elimination of homogeneous mineral acids makes
T B Adams et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 35(8), 739-751 (1997-08-01)
The Expert Panel of the Flavor and Extract Manufacturers' Association (FEMA) has assessed the safety of furfural for its continued use as a flavour ingredient. The safety assessment takes into account the current scientific information on exposure, metabolism, pharmacokinetics, toxicology
Jean-Paul Lange et al.
ChemSusChem, 5(1), 150-166 (2012-01-04)
Furfural offers a promising, rich platform for lignocellulosic biofuels. These include methylfuran and methyltetrahydrofuran, valerate esters, ethylfurfuryl and ethyltetrahydrofurfuryl ethers as well as various C(10)-C(15) coupling products. The various production routes are critically reviewed, and the needs for improvements are

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service