Skip to Content
Merck
All Photos(1)

Key Documents

51529

Supelco

Hentriacontane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)29CH3
CAS Number:
Molecular Weight:
436.84
Beilstein:
1709817
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

67-69 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C31H64/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

IUJAMGNYPWYUPM-UHFFFAOYSA-N

General description

Hentriacontane is a normal alkane, comprising of approximately 31 carbon atoms, which can be found in beewax.[1] It is also found to be a major component of candelilla wax, which upon organo-gelation can provide structure to solid-like fat-based foods, drug delivery systems and cosmetics.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles of Organic Chemistry (2015)
Ramiro Quintanilla-Licea et al.
Molecules (Basel, Switzerland), 17(9), 11229-11241 (2012-09-22)
Bioassay-guided fractionation of hexane extracts of Gymnosperma glutinosum (Asteraceae) leaves, collected in North Mexico, afforded the known compounds hentriacontane (1) and (+)-13S,14R,15-trihydroxy-ent-labd-7-ene (2), as well as the new ent-labdane diterpene (-)-13S,14R,15-trihydroxy-7-oxo-ent-labd-8(9)-ene (3). In addition, D-glycero-D-galactoheptitol (4) was isolated from the
G Yuan et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 23(8), 454-456 (2003-02-11)
The chemical constituents of Rosa bracteata Wendl. var. bracteata were studied for the first time. Six compounds were isolated from the fruits of the plant, and four of them have been identified as 5-hydroxymethyl furfural, 5-acetoxymethyl furfural, beta-sitosterol, oleanolic acid.
Antonio F M Oliveira et al.
Anais da Academia Brasileira de Ciencias, 75(4), 431-439 (2003-11-08)
The effects of the contents and chemical composition of the foliar epicuticular waxes of species from the caatinga (Aspidosperma pyrifolium, Capparis yco, Maytenus rigida and Ziziphus joazeiro) and cerrado (Aristolochia esperanzae, Didymopanax vinosum, Strychnos pseudoquina and Tocoyena formosa) were evaluated
[Antituberculous constituents of Gentiana rhodantha].
Y W Liu
Zhong yao tong bao (Beijing, China : 1981), 10(12), 33-34 (1985-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service