Skip to Content
Merck
All Photos(1)

Documents

51145

Supelco

L-Tryptophan

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86197
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

280-285 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1

InChI key

QIVBCDIJIAJPQS-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Amino acid precursor of serotonin and melatonin

Caution

Store under Argon

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirk E M Geurts et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(48), 18932-18939 (2013-11-29)
Adaptive decision-making involves interaction between systems regulating Pavlovian and instrumental control of behavior. Here we investigate in humans the role of serotonin in such Pavlovian-instrumental transfer in both the aversive and the appetitive domain using acute tryptophan depletion, known to
Kei Hiruma et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9589-9594 (2013-05-23)
The hypersensitive response (HR) is a type of strong immune response found in plants that is accompanied by localized cell death. However, it is unclear how HR can block a broad range of pathogens with different infective modes. In this
George Anderson et al.
Progress in neuro-psychopharmacology & biological psychiatry, 42, 101-114 (2012-08-30)
Schizophrenia and depression are two common and debilitating psychiatric conditions. Up to 61% of schizophrenic patients have comorbid clinical depression, often undiagnosed. Both share significant overlaps in underlying biological processes, which are relevant to the course and treatment of both
Heidi Barth et al.
Critical reviews in microbiology, 40(4), 360-368 (2012-11-24)
Indoleamine-2,3-dioxygenase (IDO) is an enzyme that catabolises tryptophan - an essential amino acid critical for T cell proliferation. Initially recognized as a first line of host defense against infectious pathogens, IDO has been subsequently identified as an important immune-regulator inhibiting
M Diksic
Journal of psychiatry & neuroscience : JPN, 26(4), 293-303 (2001-10-10)
The alpha-methyl-L-trypotophan (alpha-MTrp) method for the study of the brain serotonergic system is based on the fact that labelled alpha-MTrp is taken up by and, in part, retained in the brain, and this retention (trapping) is proportional to brain serotonin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service