Skip to Content
Merck
All Photos(4)

Documents

G4753

Sigma-Aldrich

Griseofulvin

from Penicillium griseofulvum, 97.0-102.0%

Synonym(s):

(2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
Beilstein:
95226
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Penicillium griseofulvum

Assay

97.0-102.0%

form

powder

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

ClC1=C(O[C@@]2(C(OC)=CC(C[C@H]2C)=O)C3=O)C3=C(OC)C=C1OC

InChI

1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1

InChI key

DDUHZTYCFQRHIY-RBHXEPJQSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antifungal. Mode of action: Disrupts the mitotic spindle structure and inhibits nuclear division. Induces apoptosis in human tumor cell lines.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Griseofulvin inhibits fungal mitosis.
K Gull et al.
Nature, 244(5414), 292-294 (1973-07-27)
Xing Wang et al.
Molecular cancer therapeutics, 22(4), 519-528 (2023-02-09)
Extra copies of centrosomes are frequently observed in cancer cells. To survive and proliferate, cancer cells have developed strategies to cluster extra-centrosomes to form bipolar mitotic spindles. The aim of this study was to investigate whether centrosome clustering (CC) inhibition
Dulal Panda et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(28), 9878-9883 (2005-06-30)
The antifungal drug griseofulvin inhibits mitosis strongly in fungal cells and weakly in mammalian cells by affecting mitotic spindle microtubule (MT) function. Griseofulvin also blocks cell-cycle progression at G(2)/M and induces apoptosis in human tumor cell lines. Despite extensive study
Konstantinos Drosopoulos et al.
Methods in molecular biology (Clifton, N.J.), 1953, 33-42 (2019-03-27)
Cellular models for siRNA and small molecule high-throughput screening have been widely used in the last decade to identify targets for drug discovery. As an example, we present a twofold readout approach based on cell viability and multipolar phenotype. To
Marc S Raab et al.
Cancer research, 72(20), 5374-5385 (2012-09-04)
In contrast to normal cells, malignant cells are frequently aneuploid and contain multiple centrosomes. To allow for bipolar mitotic division, supernumerary centrosomes are clustered into two functional spindle poles in many cancer cells. Recently, we have shown that griseofulvin forces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service