Skip to Content
Merck
All Photos(1)

Documents

46589

Supelco

Oxacillin sodium salt monohydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H18N3O5SNa · H2O
CAS Number:
Molecular Weight:
441.43
Beilstein:
4287093
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[H]O[H].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)c3c(C)onc3-c4ccccc4)C([O-])=O

InChI

1S/C19H19N3O5S.Na.H2O/c1-9-11(12(21-27-9)10-7-5-4-6-8-10)15(23)20-13-16(24)22-14(18(25)26)19(2,3)28-17(13)22;;/h4-8,13-14,17H,1-3H3,(H,20,23)(H,25,26);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1

InChI key

ZVIYWUUZWWBNMB-VICXVTCVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam
Oxacillin is a bactericidal agent, which can be used for the treatment of infections due to penicillinase-producing staphylococci. Its mode of action involves inhibition of bacterial cell wall synthesis and thus affects bacterial penicillin-binding proteins.

Application

Oxacillin sodium salt monohydrate has been used as a reference standard in the determination of the analyte in milk samples by receptor-binding assay test.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Desk Reference of Clinical Pharmacology (1997)
Detection of antibiotics in sheep milk by receptor-binding assays.
Beltran CM, et al.
International dairy journal, 34(2), 184-189 (2014)
Bintao Cui et al.
Journal of clinical microbiology, 51(1), 9-14 (2012-10-12)
Coagulase-negative staphylococci have been identified as major causes of late-onset neonatal bacteremia in neonatal intensive care units. Sixty isolates of Staphylococcus capitis obtained from blood cultures of neonates between 2000 and 2005 were examined in this study. Biochemical analysis confirmed
Kinga Wieczorek et al.
Foodborne pathogens and disease, 9(8), 681-685 (2012-07-26)
Four hundred seventeen retail beef meat samples purchased in the eastern part of Poland during October 2009 to January 2011 were tested for the presence of Listeria monocytogenes. It was found that 81 (19.4%) of them were positive for this
Xin-Ming Liu et al.
Pharmaceutical research, 29(11), 3169-3179 (2012-06-27)
To develop novel biomineral-binding liposomes (BBL) for the prevention of orthopedic implant associated osteomyelitis. A biomineral-binding lipid, alendronate-tri(ethyleneglycol)-cholesterol conjugate (ALN-TEG-Chol), was synthesized through Cu(I)-catalyzed Huisgen 1,3-dipolar cycloaddition (a versatile click reaction). Mixing with other excipients, the new lipid was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service