Skip to Content
Merck
All Photos(1)

Key Documents

46439

Supelco

Quinoxyfen

PESTANAL®, analytical standard

Synonym(s):

(5,7-Dichloro-4-quinolyl) (4-fluorophenyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8Cl2FNO
CAS Number:
Molecular Weight:
308.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1ccc(Oc2ccnc3cc(Cl)cc(Cl)c23)cc1

InChI

1S/C15H8Cl2FNO/c16-9-7-12(17)15-13(8-9)19-6-5-14(15)20-11-3-1-10(18)2-4-11/h1-8H

InChI key

WRPIRSINYZBGPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Quinoxyfen is a fungicide, which belongs to the family of quinolones. It finds applications in controlling powdery mildew in plants and its mode of action involves the interference with germination and/or appressorium formation.

Application

Quinoxyfen may be used as a reference standard for the determination of quinoxyfen pesticide residues in grapes by mass spectrometry coupled with gas and/or liquid chromatography.
Quinoxyfen may be used as a reference standard in the enzyme-linked immunosorbent assay developed to determine quinoxyfen in water samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F - closed cup - (External MSDS)

Flash Point(C)

> 100 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J P Zubrod et al.
Aquatic toxicology (Amsterdam, Netherlands), 169, 105-112 (2015-11-02)
Animals involved in leaf litter breakdown (i.e., shredders) play a central role in detritus-based stream food webs, while their fitness and functioning can be impaired by anthropogenic stressors. Particularly fungicides can affect shredders via both waterborne exposure and their diet
Surface-water exposure to quinoxyfen: Assessment in landscape vineyards.
Merli A, et al.
Journal of Hydrology, 383(1-2), 62-72 (2010)
Brent Warneke et al.
Plant disease, 104(4), 1167-1174 (2020-02-14)
Grape powdery mildew (GPM) fungicide programs consist of 5 to 15 applications, depending on region or market, in an attempt to achieve the high fruit quality standards demanded by the market. Understanding how fungicides redistribute and targeting redistributing fungicide to
Fate of quinoxyfen residues in grapes, wine, and their processing products.
Cabras P, et al.
Journal of Agricultural and Food Chemistry, 48(12), 6128-6131 (2000)
Immunochemical rapid determination of quinoxyfen, a priority hazardous pollutant.
Duncan H, et al.
Chemosphere (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service