Skip to Content
Merck
All Photos(1)

Key Documents

32057

Supelco

Folpet

PESTANAL®, analytical standard

Synonym(s):

2-(Trichloromethylsulfanyl)isoindole-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Cl3NO2S
CAS Number:
Molecular Weight:
296.56
Beilstein:
193373
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC(Cl)(Cl)SN1C(=O)c2ccccc2C1=O

InChI

1S/C9H4Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-4H

InChI key

HKIOYBQGHSTUDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Folpet is a contact fungicide used for the control of foliar and seed-borne diseases. Folpet is known to inhibit various oxidative enzymes, carboxylases and also enzymes required for phosphate metabolism and citrate synthesis.

Application

Folpet may be used as a reference standard for the determination of the analyte in food matrices based on QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method for extraction followed by gas chromatography coupled with electron ionization and triple quadrupole mass spectrometry (GC-EI-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of folpet and phthalimide in food by gas chromatography and mass spectrometry: Overcoming potential analytical artefacts.
Huertas-Perez JF, et al.
Food Chemistry, 260, 213-220 (2018)
Mireille Canal-Raffin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 865(1-2), 106-113 (2008-03-18)
Solid-phase extractions followed by HPLC-UV/DAD methods were developed for occupational biological monitoring or forensic investigations of the fungicide folpet using its degradation products, phthalimide and phthalamic acid as plasma biomarkers. These methods show good linearity (r>0.9955), precision (CV<15%) and accuracy
Shuju Wang et al.
Se pu = Chinese journal of chromatography, 25(2), 226-229 (2007-06-22)
A method for the simultaneous determination of captan and folpet pesticide residues in apples was developed by solid-phase extraction and high performance liquid chromatography. The sample was extracted with acetonitrile and cleaned-up by a mixture of homemade sorbent and silica
Samuel M Cohen et al.
Critical reviews in toxicology, 40(6), 531-545 (2010-06-05)
A framework has been evolving for evaluation of mode of action (MOA) of rodent toxicity and carcinogenicity findings and their relevance to humans. Folpet produces duodenal glandular tumors in mice, but is not carcinogenic in rats. A wealth of information
Aurélie Berthet et al.
Analytical and bioanalytical chemistry, 400(2), 493-502 (2011-02-22)
Agricultural workers are exposed to folpet, but biomonitoring data are limited. Phthalimide (PI), phthalamic acid (PAA), and phthalic acid (PA) are the ring metabolites of this fungicide according to animal studies, but they have not yet been measured in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service