Skip to Content
Merck
All Photos(1)

Key Documents

45995

Supelco

Cartap hydrochloride

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N3O2S2 · HCl
CAS Number:
Molecular Weight:
273.80
Beilstein:
5157539
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cl[H].CN(C)C(CSC(N)=O)CSC(N)=O

InChI

1S/C7H15N3O2S2.ClH/c1-10(2)5(3-13-6(8)11)4-14-7(9)12;/h5H,3-4H2,1-2H3,(H2,8,11)(H2,9,12);1H

InChI key

MSHXTAQSSIEBQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinxia Dai et al.
Journal of chromatography. A, 1600, 148-157 (2019-05-08)
Risk assessment of cartap residue in tea should include the exposure of cartap and its metabolite due to rapid degradation of cartap into nereistoxin. Herein, a reliable method for determination of cartap and nereistoxin in tea was developed by hydrophilic
Mahdieh Emadi et al.
Journal of biochemical and molecular toxicology, 33(7), e22325-e22325 (2019-04-21)
Cartap hydrochloride is a mildly perilous insecticide known as "Padan" which is used largely in agricultural farms to control weevil and caterpillars. The over use of cartap causes harmful effects on human health. Since the blood may acts as a
Jinxia Dai et al.
Food chemistry, 314, 126165-126165 (2020-01-24)
There are few studies for risk assessment of cartap and its metabolites, although cartap is easily transformed into metabolites which could induce higher toxicity. This study aimed to investigate the dissipation pattern of cartap and its metabolites during tea planting
Wei Liu et al.
Talanta, 101, 382-387 (2012-11-20)
The residue of pesticide has posed a serious threat to human health. Fast, broad-spectrum detection methods are necessary for on-site screening of various types of pesticides. With citrate-coated Au nanoparticles (Au NPs) as colorimetric probes, a visual and spectrophotometric method
Influence of pesticide-fertilizer combination on food intake, growth, and conversion efficiencies of Oreochromis mossambicus.
V Palanivelu et al.
Bulletin of environmental contamination and toxicology, 69(6), 908-913 (2002-11-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service