Skip to Content
Merck
All Photos(1)

Key Documents

45890

Supelco

Parathion solution

100 μg/mL in cyclohexane, PESTANAL®, analytical standard

Synonym(s):

Parathion-ethyl solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14NO5PS
CAS Number:
Molecular Weight:
291.26
Beilstein:
8912188
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3

InChI key

LCCNCVORNKJIRZ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Can mild organophosphate poisoning result in myocardial infarction?
Hossein Sanaei-Zadeh
Journal of forensic and legal medicine, 19(6), 363-363 (2012-08-01)
Courtney Dunn et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 19(5), 504-509 (2012-05-19)
Therapies exist for acute organophosphate (OP) exposure but mortality rates remain high (10% to 20%). Currently, treatment focuses on reversing the resultant cholinergic excess effects through the use of atropine. Intralipid fat emulsion (IFE) has been used to treat lipophilic
Meha Jaiswal et al.
Environmental science and pollution research international, 19(6), 2055-2062 (2012-01-05)
Removal of malathion from agricultural runoff was studied using novel copper-coated chitosan nanocomposite (CuCH)-a biopolymeric waste obtained from marine industry. Synthesis and characterization of the adsorbent using different spectral techniques like Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy, energy-dispersive
Organ donation following fatal organophosphate poisoning.
Jean-Louis Mariage et al.
Transplant international : official journal of the European Society for Organ Transplantation, 25(6), e71-e72 (2012-03-27)
Abdul Shafeeuulla Khan et al.
Journal of molecular graphics & modelling, 34, 10-17 (2012-02-07)
The extreme toxicity of organophosphorus nerve agents and pesticides mandates to employ models or simulants in place of the actual compounds in the laboratory. The importance of simulants is known, however, their efficacy for direct comparison with the toxic organophosphorus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service