Skip to Content
Merck
All Photos(1)

Key Documents

45607

Supelco

Parathion

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl O-(4-nitrophenyl) phosphorothioate, Ethylparathione, Parathion-ethyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14NO5PS
CAS Number:
Molecular Weight:
291.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3

InChI key

LCCNCVORNKJIRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Inactivation of rat liver cytochrome P-450 by the suicide substrates parathion and chloramphenicol.
J Halpert et al.
Drug metabolism reviews, 12(2), 239-259 (1981-01-01)
Courtney Dunn et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 19(5), 504-509 (2012-05-19)
Therapies exist for acute organophosphate (OP) exposure but mortality rates remain high (10% to 20%). Currently, treatment focuses on reversing the resultant cholinergic excess effects through the use of atropine. Intralipid fat emulsion (IFE) has been used to treat lipophilic
Steven B Bird et al.
Toxicology, 247(2-3), 88-92 (2008-04-02)
Organophosphorus (OP) pesticides poison more than 3,000,000 people every year in the developing world, mostly through intentional self-poisoning. Advances in medical therapy for OP poisoning have lagged, and current treatment is not highly effective with mortality of up to 40%
Meha Jaiswal et al.
Environmental science and pollution research international, 19(6), 2055-2062 (2012-01-05)
Removal of malathion from agricultural runoff was studied using novel copper-coated chitosan nanocomposite (CuCH)-a biopolymeric waste obtained from marine industry. Synthesis and characterization of the adsorbent using different spectral techniques like Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy, energy-dispersive
Alejandro A Canales-Aguirre et al.
Nutritional neuroscience, 15(2), 62-69 (2012-02-16)
One of the main concerns regarding organophosphate pesticides (OP) is their possible toxic effects. Doses that do not produce acute toxicity are capable of altering the structure and biochemistry of different tissues and organs by production of reactive oxygen species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service